Anion exchange HPLC columns, PRP-X100
Toimittaja: HAMILTON BONADUZ
79668 79669 79670 79671
554-2272EA 1690 EUR
554-2272 554-2273 554-2274 554-2275
Anion exchange HPLC columns, PRP-X100
Kromatografiakolonnit
This anion column line is designed to meet the special requirements of anion analysis. These four columns cover all aspects of standard anion analysis, separation of all 8 common anions in one run. The unique design of the highly cross-linked porous-packing material, and the strong covalent bonded surface chemistry, ensure excellent durability and high separation efficiency. These columns are designed to be used with carbonate-bicarbonate buffer in any concentration or with hydroxide gradient, and many other mobile phases.
  • Analysis of common anions in aqueous solutions
  • High resolution