TSKgel® gel filtration columns, SW-type
Toimittaja: TOSOH Bioscience
0005102 0005103 0005104 0005146 0005147 0005148 0005371 0005758 0005788 0005789 0005790 0006727 0006728 0006729 0008540 0008541 0008542 0008801 0008805 0016049 0016215 0018762 0020025 0020027 0021485 0021845 0018674 0022855 0022854 0022856 0023448 0023450 0023449 0023451 0018675 0022857 0022859 0020026 0008543 0018008
554-0891EA 3540 EUR
554-0891 554-0892 554-0893 554-0894 554-0895 554-0896 554-1605 554-1606 554-0897 554-0898 554-0899 554-0900 554-0901 554-0902 554-0903 554-0904 554-0905 554-0908 554-5604 554-5605 554-5606 554-1649 554-5609 554-5610 554-5611 554-5612 554-5607 554-5614 554-5613 554-5615 554-5317 554-5319 554-5318 554-5320 554-5608 554-5616 554-5617 TOYO0020026 554-1620 TOYO0018008
TSKgel® gel filtration columns, SW-type
Kromatografiakolonnit
TSKgel® SW size exclusion columns are widely used for aqueous analysis and isolation of proteins, antibodies, enzymes, nucleic acids and other biological macromolecules.
  • Columns show high resolution and sensitivity and have excellent solvent stability with up to 100% polar organic solvents
  • The smaller particle size of the UltraSW and SWXL materials result in reduced analysis times
  • UP-SW columns are now available for UHPLC-SEC applications improving productivity and resolution
  • All SW-type columns are available in stainless steel hardware; SWXL columns are