Coated polysaccharide columns, normal phase, CHIRALCEL®
Toimittaja: CHIRAL TECHNOLOGIES
11022 12022 12025 12324 13022 13025 13325 14035 14325 14394 15022 15025 16022 16025 17035 17045 18022 18025 14225 14032 14042 14335 14345 17032 17042 17335 17345 14794 17394 14025 14045 17025 14324 11025 12325 14311 17324 17325 14337 17311 17337 12311 17435 14511 14524 14525 42325 17594 17525 17524 17523 17522 14594 14523 14522 17511 42324 42524 42594 42525 42523 42522 42511 42394 42345 42337 42335 42592 42311 15225 16224 15226 16225 16226 17224 17225 17226 17322 17323 17423 18224 18225 18226 42224 42225 42226 42322 42323 42423 42435 42445 63311 63322 63323 63324 63325 63335 63337 63345 63394 63423 63435 63445 14224 14226 14322 14323 14423 15224 42593 42595 14595 17595 42S84 14S83 14S84 63S82 17S82 17S83 17S84 42S83 42S82 63S84 63S83 14S82 14G22 14G24 14G25 14G94 14G35 14G45 17G22 17G24 17G25 17G94 17G35 17G45 63G22 63G24 63G25 63G94 63G35 63G45 42G22 42G24 42G25 42G94 42G35 42G45
554-0004EA 960 EUR
554-0004 554-0009 554-0010 554-0014 554-5703 554-1199 554-5704 554-0019 554-0026 554-0030 554-0034 554-0035 554-0037 554-0038 554-5706 554-5707 554-0051 554-0052 554-2353 554-0018 554-0020 554-0028 554-0029 554-0040 554-0041 554-0046 554-0047 554-0115 554-0117 554-0017 554-0021 554-0103 554-0025 554-0005 554-0015 554-0024 554-0043 554-0044 554-3742 554-3744 554-3745 554-5702 554-3766 554-2446 554-2449 554-2450 554-5722 554-2457 554-2456 554-2455 554-2454 554-2453 554-2451 554-2448 554-2447 554-2452 554-5721 554-5730 554-5734 554-5731 554-5729 554-5728 554-5727 554-5726 554-5725 554-5724 554-5723 554-5732 554-5720 554-2355 554-4054 554-4053 554-2356 554-4055 554-4056 554-2357 554-4057 554-4058 554-4059 554-4060 554-4070 554-2358 554-4069 554-3389 554-2360 554-3390 554-3391 554-3392 554-3393 554-3394 554-3395 554-3417 554-3418 554-3419 554-3420 554-2359 554-3421 554-3422 554-3423 554-3424 554-3425 554-3426 554-3427 554-4047 554-4048 554-4049 554-4050 554-4051 554-4052 554-5733 554-5735 554-5705 554-5708 554-4589 554-4576 554-4577 554-4602 554-4578 554-4579 554-4580 554-4588 554-4587 554-4604 554-4603 554-4575 DAIC14G22 DAIC14G24 DAIC14G25 DAIC14G94 DAIC14G35 DAIC14G45 DAIC17G22 DAIC17G24 DAIC17G25 DAIC17G94 DAIC17G35 DAIC17G45 DAIC63G22 DAIC63G24 DAIC63G25 DAIC63G94 DAIC63G35 DAIC63G45 DAIC42G22 DAIC42G24 DAIC42G25 DAIC42G94 DAIC42G35 DAIC42G45
Coated polysaccharide columns, normal phase, CHIRALCEL®
Kromatografiakolonnit
These columns are packed with Daicel’s coated polysaccharide-based chiral stationary phases (CSPs) which are manufactured by physically coating the polysaccharide derivatives onto silica matrices. Due to this manufacturing process, a choice of mobile phase components is typically narrower than with immobilised columns in order to minimise damage to the CSPs.