TLC plates, silica gel 60, unmodified layers, ADAMANT
Toimittaja: MACHEREY-NAGEL
821005 821010 821015 821020 821025 821030 821040 821050 821060 821010.200 821040.200
552-0332EA 425 EUR
552-0332 552-0333 552-0335 552-0336 552-0337 552-0338 552-0339 552-0341 552-0342 552-0334 552-0340
TLC plates, silica gel 60, unmodified layers, ADAMANT
TLC-levyt
These unmodified standard silica layers for TLC are available as glass plates with or without fluorescent indicator. High suitability for trace analyses resulting from a UV indicator with increased brilliance and a low-noise background of the layer.
  • Efficient hardness and abrasion resistance due to an optimised binder system
  • Increased separation efficiency due to an optimised particle size distribution
Silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 5 to 17 μm.