Rat IgG2b Isotype Control [clone: LTF-2] (PE (Phycoerythrin)-Cy5®)

Toimittaja: Tonbo Biosciences
55-4031-U100
55-4031-U100EA 118 EUR
55-4031-U100
Rat IgG2b Isotype Control [clone: LTF-2] (PE (Phycoerythrin)-Cy5®)
Vasta-aineet
The LTF-2 immunoglobulin is useful as an isotype-matched control. The LTF-2 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2b antibodies.

Type: Control
Antigen: LTF-2
Clonality:
Clone: LTF-2
Conjugation: PE (Phycoerythrin)-Cy5
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR