Syringes for Shimadzu GC autosamplers
Toimittaja: HAMILTON BONADUZ
202630 202640 202643 93898-01
549-1113EA 238 EUR
549-1113 549-1114 549-1058 549-1060
Syringes for Shimadzu GC autosamplers
Ruiskut Ruiskut kromatografiaan
These syringes are used for GC autosamplers.
  • Removable needle
  • Cemented needle termination with metal flange and glass flange