Kromatografian ruiskut, micro extraction by packed sorbent (MEPS™)
Toimittaja: Trajan Scientific and Medical
006292 005291 006291 005292 006293 2928500 2928600
554-1460EA 241 EUR
554-1460 554-1454 554-1459 549-0733 549-0734 SGEA2928500 SGEA2928600
Kromatografian ruiskut, micro extraction by packed sorbent (MEPS™)
Ruiskut Ruiskut kromatografiaan
Micro Extraction by Packed Sorbent (MEPS™) is a new development in the field of sample preparation by SPE (solid phase extraction). The MEPS™ product consists of two parts, the MEPS™ syringe and the MEPS™ Barrel Insert and Needle assembly (BIN). The MEPS™ approach to sample preparation is suitable for reversed phases, normal phases, mixed mode and ion exchange chemistries. Because MEPS™ BINs can be used for sample volumes as small as 10 µl it makes them particularly well suited to on-line use with LC-MS analysis of volume limited samples.
  • The BIN contains the SPE phase that is built