HyperSep™ C18 SPE columns
Toimittaja: Thermo Fisher Scientific
60108-703 60108-302 60108-303 60108-301 60108-305 60108-304 60108-390 60108-701 60108-702 60108-376 60108-765 60108-786
549-0693EA 0 EUR
549-0693 549-0686 549-0687 549-0685 549-0689 549-0688 549-0690 549-0691 549-0692 HYPE60108-376 HYPE60108-765 HYPE60108-786
HyperSep™ C18 SPE columns
Kromatografiakolonnit
These columns feature a highly retentive alkyl-bonded phase for non polar to moderately polar compounds.
  • Hydrophobic reversed phase
  • Retains most organic analytes from aqueous matrices