VWR®, Vakiomalliset mikroruiskut
Toimittaja: VWR Collection
549-0199EA 41.2 EUR
549-0199 549-0522 549-0523 549-0524
VWR®, Vakiomalliset mikroruiskut
Ruiskut Ruiskut kromatografiaan
Standard micro syringes for GC with fixed needle.
  • All parts in contact with liquid are made of stainless steel and neutral borosilicate glass
Length in ordering table refers to needle length.