Replacement kits for plunger-in-needle syringes
Toimittaja: Trajan Scientific and Medical
033010 033011 033012 033057 033060 033605 033610 033620 033630 034055 034056 034057 034060 034610 035055 035056 035057 035058
549-0498EA 82.1 EUR
549-0498 549-0499 549-0500 549-0350 549-0501 SGEA033605 SGEA033610 SGEA033620 SGEA033630 549-0351 549-0510 549-0352 549-0512 549-0513 549-0353 549-0516 549-0354 549-0517
Replacement kits for plunger-in-needle syringes
Ruiskuneulat
Kits are available for 0,5 µl to 5,0 µl NanoVolume syringes.
Toimitustiedot: Kits are supplied with a matched plunger and needle, both must be replaced together.