VWR® PFAS Kits, 0,7 ml PP Vials with 11 mm Snap Ring Cap, Silicone/Polypropylene
Toimittaja: VWR Collection
548-7004AEA 422 EUR
548-7004A 548-7007A
VWR® PFAS Kits, 0,7 ml PP Vials with 11 mm Snap Ring Cap, Silicone/Polypropylene
Näytevialit ja pienet näyteastiat
PFAS stands for per- and polyfluoroalkyl substances, and are colloquially known as ‘forever chemicals’. Available with or without a pre-slit septum.
  • Vial type: Chromatography vial
  • Application: HPLC
  • Non certified
It is essential during the PFAS sample process to provide vials and caps that is PFAS free. Using the new PFAS vials and caps of VWR ensure the integrity of the data collected and meets all required collection standards. VWR has created a series of vials and caps that are compatible with all common autosamplers. These vials and caps