VWR®, Septa 18 mm
Toimittaja: VWR Collection
548-3223AEA 681 EUR
548-3223A 548-0806A 548-0866A 548-1464A 548-0867A 548-3224A 548-3221A 548-3315A
VWR®, Septa 18 mm
Septumi
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available
Septa choice depends on the application.