Glass crimp top vials, 2 ml, level 2 high-throughput applications, SureSTART™
Toimittaja: Thermo Fisher Scientific
6ACV11-1P 6ACV11-1PF 6ACV11-1PT 6ACV11-2P 6ACV11-2PF 6ACV11-2PT
548-2709EA 15.8 EUR
548-2709 548-2710 548-2711 548-2712 548-2713 548-2714
Glass crimp top vials, 2 ml, level 2 high-throughput applications, SureSTART™
Näytevialit ja pienet näyteastiat
Choose Thermo Scientific™ SureSTART™ 2 ml glass crimp top vials, performance level 2, to ensure high quality data with an uninterrupted workflow in high-throughput applications using GC, HPLC/UHPLC, and single or triple quadrupole MS systems.
  • Available in clear and amber glass
  • Meet all requirements of US, EU, JPN Pharmacopeia
  • Use 11 mm crimp caps and septa to seal these vials
  • Use manual or electronic crimpers and decrimpers to seal and remove the crimp caps
  • Buy in bulk for greater savings
Use these vials for routine QA/QC applications, where theMore Product Information