EZ-fill® Vials, Stevanato Group
Toimittaja: DWK Life Sciences
06B2020.6461 06B2526.7082 P05B2526.6434 P05B2526.6492 05B2060.6924 P06B2001.6461 06B2543.7082 P05B2003.6434 05B2003.6492 05B2536.6924
548-2619EA 11050 EUR
548-2619 548-2621 548-2622 548-2623 548-2624 548-2625 548-2626 548-2627 548-2628 548-2629
EZ-fill® Vials, Stevanato Group
Näytevialit ja pienet näyteastiat
EZ-fill® Vials, offered in nest and tub and trays.
  • Clean, depyrogenated, and sterilised ready-to-use (RTU) vials designed for pharmaceutical and contract manufacturing entities performing small to large batch aseptic parenteral packaging
  • ISO 8362-1 2R, 4R, 6R, 10R, and 20R vials, manufactured from borosilicate type 1, 51 expansion glass, with European Blowback
  • Nested configurations are designed for direct entry into flex fill, robotic, and combi filling operations
  • Tray configurations are suitable for small batch and in-line filling operations
  • Reduces total