VWR®, Shell Vials
Toimittaja: VWR Collection
548-0449AEA 342 EUR
548-0449A 548-2481A 548-0046A 548-0045A 548-0352A 548-0042A 548-0347A 548-0353A 548-0043A 548-0048A 548-0049A
VWR®, Shell Vials
Näytevialit ja pienet näyteastiat
Shell vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a PE plug featuring a starburst top for easier needle penetration.
  • Economical choice for many HPLC applications
  • Used in combination with PE Plugs
  • Quick assembly
Shell vials constructed from type I borosilicate glass with low metal content. Vials are available in three different sizes (1, 2 and 4 ml) in amber or clear version with a flat bottom to ensure compatibility with inserts.