Storage boxes for vials, M-T Vial File®, WHEATON®
Toimittaja: DWK Life Sciences
228780 228778 228783 228785 W228787 W228789 W228790 W228792 W228786 W228788 W228782 W228781
548-2206EA 346 EUR
548-2206 548-2205 548-2207 548-2208 548-1085 548-1084 548-1089 548-1088 548-1087 548-1086 WHEAW228782 WHEAW228781
Storage boxes for vials, M-T Vial File®, WHEATON®
Laatikot Laatikot vials
Clear plastic hinged case for convenient storage and transportation of sample vials.
  • Polycoated partitions protect vials from abrasion and breakage
  • Alphanumeric index card provides for easy vial identification
Tilaustiedot: Extra partitions and index cards can be purchased separately.
Toimitustiedot: Comes with polystyrene case, partitions, foam inserts and indexing card.