VWR®, Crimp Neck Headspace Vials ND20
Toimittaja: VWR Collection
548-0611AEA 375 EUR
548-0611A 548-0612A 548-0057A 548-0610A 548-0614A 548-0613A 548-0091A 548-2085A 548-0090A 548-0133A 548-0055A 548-0151A 548-0799A 548-0798A 548-0891A 548-1994A 548-2092A 548-2365A 548-2398A 548-1990A 548-1991A
VWR®, Crimp Neck Headspace Vials ND20
Näytevialit ja pienet näyteastiat
Headspace ND20 vials with crimp neck manufactured from 1st hydrolytic class borosilicate glass in both flat bottom and round bottom style. They can be purchased with a variety of aluminium seal styles and septa materials.
  • High performance caps/septa
  • Available with flat DIN crimp neck for tighter seal
  • Thicker wall 1,2 mm withstand higher internal pressure
Headspace ND20 vials are available in three different sizes (5, 10 and 20 ml) in amber or clear version. For use in a broad variety of instrument platforms the vials are available with round or aMore Product Information