Headspace HDHT syringes for PAL COMBI–xt® headspace autosamplers
Toimittaja: HAMILTON BONADUZ
209681 209683 209682 209685 209684 209686
548-1983EA 282 EUR
548-1983 548-1985 548-1984 548-1987 548-1986 548-1988
Headspace HDHT syringes for PAL COMBI–xt® headspace autosamplers
Ruiskut Ruiskut kromatografiaan
These headspace syringes feature a unique Glue-Free (GF) needle attachment in combination with a High Dynamic (HD) plunger. They are designed specifically for PAL COMBI–xt® headspace autosamplers. Conventional headspace syringe needles are attached to the syringe glass body using glue (cement). Common problems are detached needles or ghost peaks due to low chemical inertness of the cement to solvents or limited resistance to higher temperature. These problems are addressed with the Glue Free High Temperature (HDHT) headspace syringes.
  • Glue free needle attachment is chemically inert,