VWR®, Manual Crimper and Decapper
Toimittaja: VWR Collection
548-0069AEA 339 EUR
548-0069A 548-0071A 548-0073A 548-0074A 548-1376A 548-0658A 548-1485A 548-0499A 548-0504A 548-0070A 548-0072A 548-0500A 548-0602A 548-0605A 548-0503A 548-0655A 548-0645A 548-3288A 548-1378A 548-1374A 548-1375A 548-1377A 548-1379A 548-0646A 548-2168A 548-2169A 548-0643A 548-1486A 548-0888A
VWR®, Manual Crimper and Decapper
Korkinsulkijat ja -poistajat
Crimpers and decappers are an ideal supplement to the headspace and crimp neck vials.
  • Chemically resistant surface
  • Easy handling
  • Long life (hardened crimping jaws)
  • Adjustable crimping pressure
The crimpers and decappers allow for easy manual attachment and removal of aluminium seals to crimp neck and headspace vials.