VWR®, Septa 16 mm
Toimittaja: VWR Collection
548-0532AEA 59 EUR
548-0532A 548-0795A 548-0539A 548-0540A 548-0531A 548-0534A 548-1063A 548-0864A 548-0538A 548-1832A 548-2358A 548-0537A 548-0533A
VWR®, Septa 16 mm
Septumi
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available
Septa choice depends on the application.