VWR®, Screw Neck Vials ND13
Toimittaja: VWR Collection
548-0509AEA 356 EUR
548-0509A 548-0523A 548-0529A 548-0528A 548-0519A 548-0524A 548-0521A 548-0526A 548-0522A 548-0527A 548-0051A 548-0052A 548-0510A 548-1395A 548-3252A 548-1454A 548-3314A
VWR®, Screw Neck Vials ND13
Näytevialit ja pienet näyteastiat
ND13 screw neck vials are manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polypropylene seal styles and septa materials.
  • High performance caps/septa
  • Good handling with gloves
  • Quick assembly
ND13 screw neck vials are constructed from type I borosilicate glass with low metal content. Vials are available in amber or clear versions with a flat bottom to ensure compatibility with inserts. Associated screw caps, mostly available with assembled septa, are engineered to prevent sampleMore Product Information