VWR®, Crimp Caps ND8
Toimittaja: VWR Collection
548-3290AEA 240 EUR
548-3290A 548-3291A 548-0810A 548-0325A 548-0041A 548-0374A 548-0040A 548-0324A 548-0326A 548-0328A 548-0038A 548-0813A
VWR®, Crimp Caps ND8
Tulpat ja korkit Kromatografian näytekuppien tulpat
Crimp Caps ND8 can be purchased with a variety of septa materials. Furthermore, push-on snap caps for crimp neck vials ND8 are available.
  • High performance caps/septa
  • Fits with standard ND8 Crimp neck vials
Crimp Caps ND8 can be purchased with a variety of septa materials. Crimp caps mostly available with assembled stopper or septa are engineered to prevent sample contamination while providing consistent, secure seals.