VWR®, Septa ND9
Toimittaja: VWR Collection
548-3323AEA 49.6 EUR
548-3323A 548-3324A 548-3293A 548-3325A 548-3326A 548-1564A 548-0182A 548-0183A 548-2515A
VWR®, Septa ND9
Septumi
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available
Septa choice depends on the application.