Metyyli-tert-butyylieetteri ≥99.0% (by GC, corrected for water content), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Toimittaja: Avantor Performance Materials
Vaara

Synonyms: 2-Metoksi-2-metyylipropaani, MTBE

5398-25EA 149.97 EUR
5398-25
Metyyli-tert-butyylieetteri ≥99.0% (by GC, corrected for water content), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Metyyli-tert-butyylieetteri
Kaava: (CH₃)₃COCH₃
MW: 88.15 g/mol
Kiehumispiste: 55.2 °C
Sulamispiste: -108.6
Tiheys: 0.74
Leimahduspiste: –30 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008812
CAS-numero: 1634-04-4
UN: 2398
ADR: 3
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

For Laboratory,Research,or Manufacturing Use ≥99.0%
ACS - Assay (by GC, corrected for water) ≥99.0 %
ACS - Color (APHA) ≤10
ACS - Peroxide (as H₂O₂) ≤1 ppm
ACS - Residue after Evaporation ≤2 ppm
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.05 %
Optical Absorbance (1-cm path vs water) - 210 nm ≤1.00
Optical Absorbance (1-cm path vs water) - 225 nm ≤0.50
Optical Absorbance (1-cm path vs water) - 250 nm ≤0.10
Optical Absorbance (1-cm path vs water) - 300 nm ≤0.005
Optical Absorbance (1-cm path vs water) - 350 nm ≤0.005
Optical Absorbance (1-cm path vs water) - 400 nm ≤0.005
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR