Supelco® Ascentis® Express HILIC HPLC and UHPLC Columns
Toimittaja: Merck
53972-U 53933-U 53934-U 53938-U 53939-U 53946-U 53964-U 53967-U 53969-U 53970-U 53974-U 53975-U 53977-U 53979-U 53981-U 53520-U 53521-U 53523-U 50255-U 50256-U 50257-U 50258-U 50260-U 50261-U 50262-U 50264-U 50265-U 50268-U 50269-U 50270-U 50276-U 50278-U 50284-U 50286-U 50288-U 50289-U 50294-U 50295-U 50297-U 50298-U 51403-U 51404-U 51406-U 51408-U 51409-U 51418-U 51419-U 51421-U 51424-U 51428-U 51429-U 51430-U 51433-U
53972-UEA 928 EUR
53972-U 53933-U 53934-U 53938-U 53939-U 53946-U 53964-U 53967-U 53969-U 53970-U 53974-U 53975-U 53977-U 53979-U 53981-U 53520-U 53521-U 53523-U 50255-U 50256-U 50257-U 50258-U 50260-U 50261-U 50262-U 50264-U 50265-U 50268-U 50269-U 50270-U 50276-U 50278-U 50284-U 50286-U 50288-U 50289-U 50294-U 50295-U 50297-U 50298-U 51403-U 51404-U 51406-U 51408-U 51409-U 51418-U 51419-U 51421-U 51424-U 51428-U 51429-U 51430-U 51433-U
Supelco® Ascentis® Express HILIC HPLC and UHPLC Columns
Kromatografiakolonnit
Ascentis® Express HILIC high-speed, high-performance liquid chromatography columns are based on Fused-Core® particle design. The Fused-core particle provides a thin porous shell of high-purity silica surrounding a solid silica core.
This particle design exhibits very high column efficiency due to the shorter diffusion paths in the 0,6 µm thick porous shell around.
The bare silica stationary phase of Ascentis Express HILIC provides a column that can be used for traditional normal phase separations using non-polar, totally organic mobile phases or for aqueous normal phase chromatographyMore Product Information