1-Butanoli ≥99.9%

Toimittaja: Honeywell Chemicals
Vaara

537993-1L 537993-20L 537993-4X2.5L 537993-6X1L 537993-2.5L
537993-1LEA 57.5 EUR
537993-1L 537993-20L 537993-4X2.5L 537993-6X1L 537993-2.5L
1-Butanoli ≥99.9%
1-Butanoli

Beilstein Number: 969148
RTECS Number: EO1400000
UNSPSC Code: 12191601

Kaava: H₃C(CH₂)₂CH₂OH
Kiehumispiste: 119 °C (1,013 hPa)
Sulamispiste: <-90 °C
Tiheys: 0,81 g/cm³ (20 °C)
Leimahduspiste: 9,4 %(V)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002964
CAS-numero: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Identity (IR) complying
Assay (GC) Min. 99.80 %
Density (D 20/4) 0.809 - 0.811
Non-volatile matter Max. 20 mg/l
Water (Karl Fischer) Max. 0.1 %
Free acid (as CH₃COOH) Max. 0.005 %
Iron (Fe) Max. 0.3 ppm
Di-n-butyl ether (GC) Max. 0.100 %
Isobutyl alcohol (GC) Max. 0.100 %
n-Butyraldehyde (GC) Max. 0.020 %
Carbonyl compounds (as CO) Max. 0.050 %
APHA Max. 10
Autoignition Temperature 649 °F
Refractive Index n20/D 1.399 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR