Multi anion standard 2 for IC, TraceCERT®, Supelco®
Toimittaja: Merck
53798-100ML
53798-100MLEA 166 EUR
53798-100ML
Multi anion standard 2 for IC, TraceCERT®, Supelco®
Standardiaineet ja vertailumateriaalit Anionic Standards Multi-Element Anioinic Standards
Sertifikaatit: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.