VWR® Soxhlet Extractors
Toimittaja: VWR Collection
537-0047EA 61.5 EUR
537-0047 537-0048 537-0049 537-0050 537-0045 537-0046
VWR® Soxhlet Extractors
Uuttolaitteet ja uutoshylsyt Soxhlet- ja Soxhlet-tyypin uuttolaitteet
Borosilicate 3.3 glass.
  • High chemical resistance
  • High temperature resistance and high resistance to thermal shock
  • Standard ground socket
For the extraction from solids with solvents. The solvent is recirculated continuously and distilled from the extracted sample before contacting the solid again. The extract is concentrated in a distillation flask.
Tilaustiedot: The distillation flask and condenser must be ordered separately.