Supelco® Ascentis® Express PAH HPLC Columns
Toimittaja: Merck
53551-U 53555-U 53556-U 53513-U 53532-U 53533-U 53534-U 53535-U 53538-U 53539-U 53540-U 53541-U 53550-U
53551-UEA 547 EUR
53551-U 53555-U 53556-U 53513-U 53532-U 53533-U 53534-U 53535-U 53538-U 53539-U 53540-U 53541-U 53550-U
Supelco® Ascentis® Express PAH HPLC Columns
Kromatografiakolonnit
Ascentis® Express PAH is a non endcapped C18 bonded phase with a proprietary manufacturing process designed on proven Fused-Core® technology to provide a fast and efficient separation of PAH compounds.
  • 2,7 µm Fused-Core® particles provide reliable and highly efficient separations and lower column back pressure compared to sub 2 µm particles
  • Excellent suitability for UV, fluorescence and MS detection
  • Application-related lot analysis and single column performance testing
Ascentis® Express PAH is a high speed, high performance liquid chromatographyMore Product Information