Büchner funnels, Quickfit®
Toimittaja: SciLabware
SF3A33 SF3A32 SF3B33
532-0097EA 137 EUR
532-0097 201-8681 201-8682
Büchner funnels, Quickfit®
Suppilot Suodatussuppilot Büchner-suppilot
PYREX®, borosilikaattilasia.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With sintered glass disc and integral ground glass cone.