Nikkelidibromidi, vedetön 99%

Toimittaja: Thermo Fisher Scientific
Vaara

53115.18 53115.30
53115.18EA 271 EUR
53115.18 53115.30
Nikkelidibromidi, vedetön 99%
Nikkelidibromidi

Purity: 99% CAS No.: 13462-88-9 Molecular Formula: Br2Ni Formula Weight: 218.53 Physical Form: Crystalline Density: 5.098 Melting Point: 963° Merck Reference: 14,6502 UN No.: 3077 MDL No.: MFCD00011141

Kaava: Br₂Ni
Sulamispiste: 963 °C
Tiheys: 5,098
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011141
CAS-numero: 13462-88-9
EINECS: 236-665-0
UN: 3077
ADR: 9,III
Merck Index: 14,6502

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR