Rotational viscometers, ROTAVISC
Toimittaja: IKA
10013828 10013829 10013826 10013827
531-1489EA 3820 EUR
531-1489 531-1490 531-1487 531-1488
Rotational viscometers, ROTAVISC
Viskosimetrit Rotaatioviskosimetrit
The ROTAVISC series determines the viscosity of liquids in all areas of application from the laboratory to quality control.
  • Easy to operate, highest measurement accuracy and stepless speed
  • TFT display for an intuitive menu guidance
  • Time setting range from 0,017 to 6000 min
  • Overload protection and right side rotation
  • PT100 connection for external temperature sensor
A digital spirit level also supports the correct set-up of the start-up condition and a simple ramp function simplifies repetitive tasks.
Sertifikaatit: IP 40 protection class accordingMore Product Information