VWR® LED Digital Rotary Evaporator
Toimittaja: VWR Collection
531-1369EA 1660 EUR
531-1369 531-1370 531-1372 531-1373 531-1374
VWR® LED Digital Rotary Evaporator
Haihduttimet Pyöröhaihduttimet
This is a basic chemical laboratory instrument suitable for evaporation, distillation and separation of chemicals. It can be used as a multifunctional system in combination with a vacuum pump, vacuum controller and refrigerated circulator to provide excellent distillation solutions in many laboratory applications.
  • 5 L heating bath with a wide temperature range (ambient to 180 °C), independent temperature control allows it to be used separately. Water/oil heating mode can only be changed via a switch
  • Manual lift combined with auxiliary lift for precise