Mikrosentrifugiputket, mitta-asteikolliset, turvakorkit
Toimittaja: Thermo Fisher Scientific
3454 3455-11 3456 3457 3457IW 3455IW
525-1504EA 36.9 EUR
525-1504 525-1505 525-1506 525-1507 525-1510 525-1519
Mikrosentrifugiputket, mitta-asteikolliset, turvakorkit
Putket Mikrosentrifugiputket
Manufactured from virgin polypropylene these microcentrifuge tubes ensure sample integrity and security during boiling, freezing, incubation and centrifugation.
  • Hinged locking ball reduces stress on the thumb
  • Smooth locking surface reduces the possibility of ripped or torn gloves
  • Graduation marks on the microcentrifuge tube allow for effective sample reference
  • Sterile product is certified free of RNase, DNase, DNA, ATP, bioburden and endotoxins
  • Nonsterile product is certified free of RNase, DNase, DNA and pyrogens
Exceptionally stable performanceMore Product Information