VWR®, Centrifuge Tubes
Toimittaja: VWR Collection
525-0638EA 91.2 EUR
525-0638 525-0639 525-0640
VWR®, Centrifuge Tubes
Putket Sentrifugiputket
PS, clear, conical, with flat cap with integral sealing.
  • Electron beam irradiated
  • Longer length screw caps with sealing ring prevent any leakage
  • Engraved graduations at the conical bottom of each tube
  • Graduations and writing area are chloroform-resistant
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters