Sentrifugit, pöytämalli, ilmajäähdytteinen/jäähdyttävä, 5810 / 5810 R (General Lab Product)
Toimittaja: EPPENDORF
5810000.010 5810000.320 5810000.420 5810000.525 5810000.620 5811000.015 5811000.325 5811000.420 5811000.620 5811000.820
521-2428EA 6100 EUR
521-2428 521-2640 521-2642 521-2644 521-3508 521-3509 521-2641 521-2643 521-2645 EPPE5811000.820
Sentrifugit, pöytämalli, ilmajäähdytteinen/jäähdyttävä, 5810 / 5810 R (General Lab Product)
Sentrifugit Pöytämalliset sentrifugit
This reliable centrifuge for medium to high-throughput applications combines extraordinary versatility and capacity for both tubes and plates with an extraordinary compact footprint. Bucket adapters and adapter modules can be stacked until the required tube height is attained and then lowered onto the holders in the rectangular buckets. The stable plastic adapters are robustly constructed and can be easily cleaned, even autoclaved in their entirety in the event of contamination with biologically active sample material.
  • Automaattinen roottorintunnistus takaa turvallisuuden tunnistamalla