Centrifuges, bench top, refrigerated, 5910Ri (general lab product)
Toimittaja: EPPENDORF
5943000.011 5943000.149 5943000.173 5943000.216 5943000.316 5943000.516 5943000.904
521-3382EA 12170 EUR
521-3382 521-3386 521-3388 521-3391 521-3394 521-3399 521-3402
Centrifuges, bench top, refrigerated, 5910Ri (general lab product)
Sentrifugit Pöytämalliset sentrifugit
The refrigerated benchtop centrifuge 5910Ri stands out for its ease of use and versatility. With a range of innovative solutions in usability, documentation and user management. The centrifuge has designed to accommodate a particularly broad spectrum of applications. Cell harvests in bottles with capacities up to 1000 ml, large-scale DNA and RNA isolations as well as Ficoll® gradients for the purification of lymphocytes and monocytes are merely a few examples of the many areas of application.
  • 7" VisioNize® touchscreen interface with an intuitive user experience provides fast and error-