Centrifuges, Multifuge™ X4/X4R Pro
Toimittaja: Thermo Scientific
75009900 75009915 75016066 75016067 75016077 75016087
521-2728EA 6608 EUR
521-2728 521-2730 521-2735 521-2736 521-2739 521-3499
Centrifuges, Multifuge™ X4/X4R Pro
Sentrifugit Pöytämalliset sentrifugit
Process a range of general purpose applications including cell culture, microplate, bioproduction, blood separation and microbiology with the Thermo Scientific™ Multifuge™ X4 Pro centrifuge series featuring new touch screen technology, making daily tasks easier.
  • Process up to 40×50 ml conical tubes, 196×5/7 ml blood tubes, 24 micro plates
  • Process clinical protocols, cell culture applications and microplate processing
  • Modern, intuitive touch screen interface enables easy access and programming for faster results
  • Lighthouse mode: Easily monitor run progress