VWR® Mega Star 4.0 / 4.0R, Centrifuges, Bench Top, Ventilated/Refrigerated,
Toimittaja: VWR Collection
521-2662EA 6550 EUR
521-2662 521-2663 521-2664 521-2665
VWR® Mega Star 4.0 / 4.0R, Centrifuges, Bench Top, Ventilated/Refrigerated,
Sentrifugit Pöytämalliset sentrifugit
These versatile centrifuges are ideal for a range of routine applications in pharmaceutical and biotechnology industries, as well as for academic research. They offer exceptional capacity and an ergonomic design which facilitates work on a bench top.
  • Auto-Lock® III rotor system: Tool-free rotor exchange system enables quick rotor exchange; with just the push of a button users can quickly change rotors and easily access the rotor chamber for cleaning
  • Aerosol-tight ClickSeal® bucket sealing system: Glove friendly one-handed open/close capability, no need for