Lisävarusteet, Medifuge™ centrifuge
Toimittaja: Thermo Scientific
75008818 75008813 75008815 75008817 75008810 75008816
521-2233EA 73.3 EUR
521-2233 521-2229 521-2230 521-2232 521-2228 521-2231
Lisävarusteet, Medifuge™ centrifuge
Sentrifugit Pöytämalliset sentrifugit
Run common clinical IVD blood tubes, gel tubes, 10 ml standard syringes and 15 ml conical tubes with the Thermo Scientific™ DualSpin™ rotor. This lightweight eight-place rotor, used with the Thermo Scientific™ Medifuge™ small benchtop centrifuge, features a unique hybrid design with interchangeable fixed angle and swinging buckets. It can accommodate 1,4 to 15 ml tubes with two spacer options. You can even run both fixed angle and swinging buckets at the same time to compare separation performance.
  • Unique DualSpin rotor, features hybrid design with interchangeable fixed angle and