Sentrifugit, pöytämalli, ilmajäähdytteinen/jäähdyttävä, 5810 / 5810 R (IVD)
Toimittaja: EPPENDORF
5810000.017 5811000.010 5810000.068 5811000.061 5810000.327 5811000.320 5810000.424 5811000.428 5810000.360 5811000.363 5810000.467 5811000.460 5810000.521 5811000.622 5811000.665 5811000.827 5810000.629
521-0076EA 6400 EUR
521-0076 521-0103 521-0143 521-0148 521-0996 521-0998 521-0997 521-0999 521-1144 521-1145 521-1146 521-1147 521-1716 521-1684 EPPE5811000.665 521-2054 521-2053
Sentrifugit, pöytämalli, ilmajäähdytteinen/jäähdyttävä, 5810 / 5810 R (IVD)
Sentrifugit Pöytämalliset sentrifugit
This reliable centrifuge for medium to high-throughput applications combines extraordinary versatility and capacity for both tubes and plates with an extraordinary compact footprint. Bucket adapters and adapter modules can be stacked until the required tube height is attained and then lowered onto the holders in the rectangular buckets. The stable plastic adapters are robustly constructed and can be easily cleaned, even autoclaved in their entirety in the event of contamination with biologically active sample material.
  • Automatic rotor recognition senses rotor type and sets maximum