n-Heptaani ≥99.0% (by GC, corrected for water content), AR®, Macron Fine Chemicals™

Toimittaja: Avantor Performance Materials
Vaara

5177-45EA 882.47 EUR
5177-45 5177-25 5177-26
n-Heptaani ≥99.0% (by GC, corrected for water content), AR®, Macron Fine Chemicals™
n-Heptaani
Kaava: H₃C(CH₂)₅CH₃
Kiehumispiste: 98,4 °C (1013 hPa)
Tiheys: 0,6838 g/cm³ (20 °C)
Leimahduspiste: –4 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009544
CAS-numero: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

For Laboratory,Research,or Manufacturing Use ≥99.0%
Assay (CH₃(CH₂)₅CH₃) (by GC, corrected for water) ≥99.0 %
Acidity (as CH₃COOH) Passes Test
Residue after Evaporation ≤0.001 %
Sulfur Compounds (as S) ≤0.005 %
Specific Gravity at 60°/60°F 0.68 - 0.72
Thiophene Passes Test
Appearance (clear, colorless, volatile liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR