Suodatinsuppilo MicroFunnel™
Toimittaja: Cytiva (Formerly Pall Lab)
4800 4803 4804 4805 4806 4810 4815 4817 4818 4801 4828 4852 4800ME 4810ME 4815ME 4801ME 4804ME
516-8958EA 427 EUR
516-8958 516-8960 515-0050 516-8962 516-8963 516-8965 516-8966 516-8968 516-8969 515-0048 515-0158 516-0427 516-1228 516-1234 516-1235 516-1229 516-1233
Suodatinsuppilo MicroFunnel™
Suppilot Membraanisuodatinsuppilot
Save time with disposable units by eliminating the cleaning and sterilisation required with reusable funnels.
  • Disposable
  • Ideal for aqueous solutions
  • Squeeze separation
MicroFunnel™ filter funnels are ideal for testing any aqueous solution for microbial contamination using the principles of Membrane Filter (MF) technique.
The unique squeeze separation makes membrane retrieval easy. Choose from 0,45 µm GN-6 Metricel® (mixed cellulose ester) membrane used for most applications, 0,2 and 0,45 µm Supor® (polyethersulfone) membrane to minimise binding ofMore Product Information