Absorbent pad kits
Toimittaja: Cytiva (Formerly Pall Lab)
66025 66190
516-9012EA 358 EUR
516-9012 516-7850
Absorbent pad kits
Pyyhkeet, liinat ja sienet Puhdistuspakkaukset ja -tyynyt
Handy dispenser kits allow one-handed dispensing of cellulose absorbent pads.
  • One-handed operation
  • Holds up to 100 pads
  • Sterile pads are sterilized by gamma irradiation
  • No hazardous EtO residuals
Dispense a clean cellulose pad into a petri dish without touching the pad.
Dispenser holds one tube of 100 absorbent pads. Tubes drop quickly into the hand dispenser for easy use. Absorbent pads are ideal for absorbing 2 ml of broth media to culture colonies in accordance with the membrane filter test method.
Pad Ø: 45.5 mm (1³/₄").
Pad thickness: 35 mil.More Product Information