Funnels for vacuum filtration
Toimittaja: Cytiva (Formerly Pall Lab)
4203 4204
516-7579EA 317 EUR
516-7579 516-7580
Funnels for vacuum filtration
Suppilot Suodatussuppilot Suodatussuppilot
Polysulfone, transparent.
  • Transparent funnels with graduations permit easy visual measurement
  • Less expensive than stainless steel and more durable than glass
  • Changing filters is easy
  • Twist-lock coupling minimises filter tearing
Ideal for liquid clarification, vacuum filtration, and scintillation counting studies.