Ruiskusuodatin, Minisart® SRP
Toimittaja: Sartorius
17558-K 17558-Q 17559-K 17559-Q 17573-Q 17574-K 17574-Q 17575-K 17575-Q 17575-S 17576-K 17576-Q 17576-S 17820-K 17820-Q 17575-ACK 17573-K 17573-ACK 17844-Q S7575-FXOSK
611-0705EA 261 EUR
611-0705 510-3244 611-0706 510-3245 512-3246 512-3147 510-3247 512-3148 512-3248 SART17575-S 512-3149 510-3249 SART17576-S 512-3142 512-3284 516-7035 512-3146 516-0905 516-0683 514-1238
Ruiskusuodatin, Minisart® SRP
Suodattimet Ruiskusuodattimet
Elimination of particles from samples prior to HPLC or other chromatographic analysis is essential in order to maintain the integrity of the chromatography column and to maximise its operating life time. Minisart® syringe filters optimised for sample preparation consist of a polypropylene (PP) housing and membrane components featuring maximum chemical compatibility and minimum extractables to ensure excellent results. Due to the typical range of volumes from less than 1 ml to 100 ml these filters are available in three different diameters with an effective filtration area of 0,12, 1,7 and 4,8More Product Information