Ultrasuodatuskasetti, tangentiaalivirtauksella, Minimate™
Toimittaja: Cytiva (Formerly Pall Lab)
OA010C12 OA030C12 OA100C12 OA001C12 OA003C12 OA050C12 OA070C12 OA300C12 OA500C12 OA990C12 OA005C12
516-6879EA 450 EUR
516-6879 516-6880 516-6881 515-0116 515-0117 515-0119 515-0120 515-0121 515-0122 515-0123 515-0118
Ultrasuodatuskasetti, tangentiaalivirtauksella, Minimate™
Suodattimet Tangenttivirtauskasetit
The disposable Minimate™ TFF capsules, with Omega™ membrane, can perform concentration processes desalting or buffer exchange on the same system with minimal user intervention, and achieves high concentration factors in a single step due to the low hold-up volume of the devices.
  • Scaleable
  • Works with a variety of positive pressure laboratory pumps
  • High product recovery
  • 100% integrity tested
The capsules will process samples of up to 1 litre, or efficiently concentrate samples as little as 5 ml.
Toimitustiedot: Supplied with all fittings and tubingMore Product Information