Qualitative filter papers, standard grades, grade 6, Whatman™
Toimittaja: Whatman products (Cytiva)
1006-070 1006-090 1006-110 1006-125 1006-150 1006-185 1006-240 1006-042
516-3606EA 19.8 EUR
516-3606 512-1036 512-1037 516-3610 516-3611 512-1038 516-0603 516-5127
Qualitative filter papers, standard grades, grade 6, Whatman™
Suodattimet Suodatinpaperit Kvalitatiivinen suodatinpaperi
A standard grade filter for boiler water analysis.
  • Twice as fast as Grade 5 with similar fine particle retention
  • Often specified for boiler water analysis applications
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.