Membrane filters, cellulose acetate, Whatman™
Toimittaja: Whatman products (Cytiva)
7000-0002 7000-0004 7001-0004 10403012 10403112 10404001 10404006 10404012 10404014 10404026 10404031 10404044 10404106 10404112 10404114 10404126 10404131 10404180 10404139
514-0769EA 231 EUR
514-0769 514-0770 514-0103 514-2322 514-0100 514-2350 514-2351 514-2352 514-2353 516-5015 514-2354 516-5016 514-2361 514-2362 514-2363 516-5018 514-2365 516-5020 516-5019
Membrane filters, cellulose acetate, Whatman™
Suodattimet Membraanisuodattimet
Cytiva's Whatman cellulose acetate membrane filters have very low protein binding, which minimizes sample loss when filtering protein-based aqueous samples.
  • Pure cellulose acetate composition, minimising sample contamination
  • Hydrophilic, for use with aqueous and alcoholic media
  • High solvent resistance, particularly to low molecular weight alcohols
  • High physical strength and a heat resistance up to 180 °C
  • Can be sterilized by most common methods without sacrificing membrane integrity
Due to their high chemical resistance, cellulose acetate membraneMore Product Information