Syringe filters, sterile, Whatman™ Anotop
Toimittaja: Whatman products (Cytiva)
6809-1112 6809-2112 6809-1102 6809-1122 6809-2102 6809-2122
516-1128EA 829 EUR
516-1128 516-1504 516-1127 516-1129 516-1502 516-1507
Syringe filters, sterile, Whatman™ Anotop
Suodattimet Ruiskusuodattimet
Whatman Anotop sterile syringe filters from Cytiva contain an inorganic Anopore membrane. The low protein binding exhibited by this sterile membrane filter make it well suited for biological filtration.
  • Sterile syringe filters for critical applications
  • Aluminum-based inorganic Anotop syringe filters in three pore sizes
  • Compatible with most organic solvents and aqueous samples
  • Low hold-up volume (< 20 μl for Anotop 10) for maximum sample recovery
  • Made without wetting agents or adhesives for low risk of sample contamination
  • Capillary pore structure